siRNA Details
| Gene Symbol: Dusp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007268.1 | NP_001007269.1 | 8 | 1415 | ||
| NM_013849.3 | NP_038877.2 | 4 | 740 | |||
| GenBank mRNA | AB027003.1 | - | 3 | 521 | ||
| AF237620.1 | - | 4 | 723 | |||
| AK006247.1 | - | 4 | 741 | |||
| AY674052.1 | - | 8 | 1415 | |||
| BC100421.1 | - | 3 | 646 | 30919819 | ||

Dusp13
Genomic Map