siRNA Details
| Gene Symbol: Zeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289521.1 | NP_001276450.1 | 3 | 580 | ||
| NM_015753.4 | NP_056568.2 | 4 | 917 | |||
| GenBank mRNA | AF033116.1 | - | 3 | 432 | ||
| AK122312.1 | - | 4 | 490 | |||
| AK141157.1 | - | 3 | 548 | |||
| BC060699.1 | - | 4 | 654 | 6408845 | ||

Zeb2
Genomic Map