siRNA Details
| Gene Symbol: Hunk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015755.2 | NP_056570.1 | 11 | 1652 | ||
| GenBank mRNA | AF055919.2 | - | 11 | 1652 | ||
| AF167987.1 | - | 11 | 1664 | |||
| AK144639.1 | - | 11 | 1635 | |||
| AK147425.1 | - | 11 | 2038 | |||
| AK147734.1 | - | 11 | 1406 | |||
| BC137788.1 | - | 11 | 1713 | 8860807 | ||
| BC137790.1 | - | 11 | 1712 | 8860809 | ||

Hunk
Genomic Map