siRNA Details
Gene Symbol: Vezf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016686.4 | NP_057895.2 | 6 | 1756 | ||
GenBank mRNA | AF104410.3 | - | 6 | 1756 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vezf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016686.4 | NP_057895.2 | 6 | 1756 | ||
GenBank mRNA | AF104410.3 | - | 6 | 1756 |