siRNA Details
| Gene Symbol: Tpmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016785.2 | NP_058065.2 | 9 | 1034 | ||
| GenBank mRNA | AF037043.1 | - | 9 | 943 | ||
| AF046887.1 | - | 9 | 1020 | |||
| AK002335.1 | - | 9 | 836 | |||
| AK079074.1 | - | 2 | 289 | |||
| BC021598.1 | - | 8 | 919 | 5098857 | ||
| Gene Symbol: Gm8479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF037046.1 | - | 1 | 1027 | ||
| AF037047.1 | - | 1 | 998 | |||

Tpmt
Genomic Map