siRNA Details
| Gene Symbol: Tcirg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136091.2 | NP_001129563.1 | 13 | 1626 | ||
| NM_001167784.1 | NP_001161256.1 | 13 | 1971 | |||
| NM_016921.3 | NP_058617.3 | 13 | 1667 | |||
| GenBank mRNA | AB022322.1 | - | 13 | 1533 | ||
| AF218253.1 | - | 13 | 1605 | |||
| AF218254.1 | - | 6 | 1288 | |||
| AK004094.1 | - | 4 | 477 | |||
| AK155140.1 | - | 13 | 1972 | |||
| BC006761.1 | - | 13 | 1667 | 3257437 | ||
| BC085234.1 | - | 13 | 1527 | 5322429 | ||

Tcirg1
Genomic Map