siRNA Details
Gene Symbol: ADGRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079858.2 | NP_001073327.1 | 10 | 620 | ||
NM_001079859.2 | NP_001073328.1 | 9 | 578 | |||
NM_001079860.2 | NP_001073329.1 | 8 | 554 | |||
NM_001184833.1 | NP_001171762.1 | 9 | 572 | |||
NM_001184834.1 | NP_001171763.1 | 10 | 620 | |||
NM_001184835.1 | NP_001171764.1 | 7 | 506 | |||
NM_001184836.1 | NP_001171765.1 | 8 | 548 | |||
NM_001184837.1 | NP_001171766.1 | 8 | 530 | |||
NM_005756.3 | NP_005747.2 | 10 | 611 | |||
XM_006724455.3 | XP_006724518.1 | 10 | 478 | |||
XM_011545434.1 | XP_011543736.1 | 10 | 443 | |||
XM_011545435.2 | XP_011543737.1 | 10 | 1173 | |||
GenBank mRNA | AF538954.1 | - | 10 | 541 | ||
AF539455.1 | - | 8 | 451 | |||
AF539456.1 | - | 8 | 475 | |||
AK090402.1 | - | 2 | 5425 | |||
AK291012.1 | - | 10 | 455 | |||
AY143364.1 | - | 7 | 427 | |||
AY143365.1 | - | 9 | 499 | |||
AY143366.1 | - | 9 | 493 | |||
AY143367.1 | - | 8 | 469 | |||
AY148343.1 | - | 10 | 541 | |||
BC099901.1 | - | 9 | 405 | 30349668 | ||
BC113978.1 | - | 8 | 373 | 40081713 | ||
BC113979.1 | - | 7 | 331 | 40081711 | ||
X81892.1 | - | 10 | 441 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
