siRNA Details
| Gene Symbol: Ube2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019668.3 | NP_062642.1 | 6 | 1087 | ||
| GenBank mRNA | AF089812.1 | - | 6 | 1093 | ||
| AF383148.1 | - | 6 | 1093 | |||
| AK133723.1 | - | 6 | 1096 | |||
| AK150417.1 | - | 6 | 1070 | |||
| AK153327.1 | - | 6 | 1070 | |||
| AK160527.1 | - | 6 | 1082 | |||
| AK164074.1 | - | 6 | 1096 | |||
| BC026053.1 | - | 6 | 1085 | 4012075 | ||

Ube2a
Genomic Map