siRNA Details
| Gene Symbol: Sh2d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025571.2 | NP_001020742.1 | 7 | 1219 | ||
| NM_021309.3 | NP_067284.1 | 7 | 1243 | |||
| GenBank mRNA | AF203343.1 | - | 7 | 1250 | ||
| AK037489.1 | - | 7 | 941 | |||
| AK172182.1 | - | 7 | 971 | |||
| BC034847.1 | - | 7 | 951 | 3327113 | ||
| U69490.1 | - | 7 | 929 | |||

Sh2d2a
Genomic Map