siRNA Details
| Gene Symbol: Dysf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077694.2 | NP_001071162.1 | 9 | 1231 | ||
| NM_021469.3 | NP_067444.2 | 9 | 1049 | |||
| GenBank mRNA | AF188290.2 | - | 8 | 824 | ||
| AK131144.1 | - | 9 | 1046 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dysf| Gene Symbol: Dysf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077694.2 | NP_001071162.1 | 9 | 1231 | ||
| NM_021469.3 | NP_067444.2 | 9 | 1049 | |||
| GenBank mRNA | AF188290.2 | - | 8 | 824 | ||
| AK131144.1 | - | 9 | 1046 | |||