siRNA Details
| Gene Symbol: Tulp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021478.1 | NP_067453.1 | 13 | 1266 | ||
| GenBank mRNA | AF085681.1 | - | 13 | 1314 | ||
| AF105711.1 | - | 13 | 1314 | |||
| AK053880.1 | - | 7 | 769 | |||
| AK164621.1 | - | 3 | 270 | |||
| BC014820.1 | - | 13 | 1272 | |||
| BC029222.1 | - | 13 | 1305 | 4504473 | ||
| BC038073.1 | - | 1 | 158 | |||

Tulp1
Genomic Map