siRNA Details
| Gene Symbol: Tlx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021901.3 | NP_068701.1 | 3 | 1754 | ||
| GenBank mRNA | BC018246.1 | - | 3 | 1754 | 4164519 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tlx1| Gene Symbol: Tlx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021901.3 | NP_068701.1 | 3 | 1754 | ||
| GenBank mRNA | BC018246.1 | - | 3 | 1754 | 4164519 | |