siRNA Details
| Gene Symbol: Rsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025290.3 | NP_079566.1 | 2 | 249 | ||
| GenBank mRNA | AB006535.1 | - | 2 | 237 | ||
| AK005739.1 | - | 2 | 201 | |||
| BC049584.1 | - | 2 | 249 | 6744132 | ||
| DQ141549.1 | - | 2 | 148 | |||
| DQ141550.1 | - | 2 | 148 | |||
| DQ141551.1 | - | 2 | 148 | |||

Rsph1
Genomic Map