siRNA Details
| Gene Symbol: Rsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025290.3 | NP_079566.1 | 4 | 416 | ||
| GenBank mRNA | AB006535.1 | - | 4 | 404 | ||
| AK005739.1 | - | 4 | 368 | |||
| BC049584.1 | - | 4 | 416 | 6744132 | ||
| DQ141549.1 | - | 4 | 315 | |||
| DQ141550.1 | - | 4 | 315 | |||
| DQ141551.1 | - | 4 | 315 | |||
| DQ141553.1 | - | 4 | 315 | |||
| DQ141554.1 | - | 4 | 315 | |||
| DQ141555.1 | - | 3 | 201 | |||
| DQ141556.1 | - | 3 | 201 | |||

Rsph1
Genomic Map