siRNA Details
| Gene Symbol: SNRPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006938.3 | NP_008869.1 | 2, 3 | 260 | ||
| GenBank mRNA | AB451227.1 | - | 1 | 99 | ||
| AB451350.1 | - | 1 | 99 | |||
| AJ577268.1 | - | 2, 3 | 192 | |||
| BC001721.1 | - | 2, 3 | 221 | 3355716 | ||
| BC072427.1 | - | 2, 3 | 147 | 6464946 | ||
| BU565530.1 | - | 2, 3 | 262 | |||
| CR542239.1 | - | 1 | 99 | |||
| HY043971.1 | - | 2, 3 | 258 | |||
| J03798.1 | - | 2, 3 | 249 | |||

SNRPD1
Genomic Map