siRNA Details
| Gene Symbol: SPTBN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006946.2 | NP_008877.1 | 2, 3 | 227 | ||
| XM_005274192.4 | XP_005274249.1 | 3, 4 | 889 | |||
| XM_005274193.3 | XP_005274250.1 | 2, 3 | 663 | |||
| XM_006718671.3 | XP_006718734.1 | 3, 4 | 289 | |||
| XM_017018174.1 | XP_016873663.1 | 3, 4 | 981 | |||
| XM_017018175.1 | XP_016873664.1 | 3, 4 | 610 | |||
| XM_017018176.1 | XP_016873665.1 | 3, 4 | 330 | |||
| XM_017018177.1 | XP_016873666.1 | 2, 3 | 519 | |||
| XM_017018178.1 | XP_016873667.1 | 2, 3 | 202 | |||
| GenBank mRNA | AB008567.1 | - | 2, 3 | 227 | ||
| BC139788.1 | - | 2, 3 | 198 | |||

SPTBN2
Genomic Map