siRNA Details
| Gene Symbol: Txnl4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025299.3 | NP_079575.1 | 2, 3 | 289 | ||
| GenBank mRNA | AK004039.1 | - | 2, 3 | 246 | ||
| AK007331.1 | - | 2, 3 | 251 | |||
| AK148287.1 | - | 2, 3 | 272 | |||
| AK166497.1 | - | 2, 3 | 265 | |||
| BC031634.1 | - | 2, 3 | 230 | 3662554 | ||
| BY065764.1 | - | 2, 3 | 290 | |||

Txnl4a
Genomic Map