siRNA Details
| Gene Symbol: Mrpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177658.1 | NP_001171129.1 | 5 | 3205 | ||
| NM_025300.4 | NP_079576.1 | 5 | 920 | |||
| GenBank mRNA | AK017824.1 | - | 5 | 883 | ||
| BC021304.1 | - | 5 | 916 | 3498041 | ||
| BC027233.1 | - | 5 | 905 | 3498065 | ||
| BC068230.1 | - | 5 | 3187 | |||

Mrpl15
Genomic Map