siRNA Details
| Gene Symbol: Deb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026794.2 | NP_081070.1 | 3 | 267 | ||
| GenBank mRNA | AK003863.1 | - | 3 | 275 | ||
| AK049702.1 | - | 2 | 2158 | |||
| AK078209.1 | - | 2 | 379 | |||
| AK085962.1 | - | 1 | 2207 | |||
| AK213061.1 | - | 2 | 157 | |||
| BC027548.1 | - | 3 | 267 | 1532650 | ||

Deb1
Genomic Map