siRNA Details
| Gene Symbol: Tfec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031198.3 | NP_112475.1 | 7 | 1633 | ||
| GenBank mRNA | AF077742.1 | - | 7 | 1633 | ||
| AK137141.1 | - | 7 | 1598 | |||
| AK156327.1 | - | 7 | 1546 | |||
| AK159579.1 | - | 7 | 1566 | |||
| BC052832.1 | - | 6 | 1521 | |||
| BC064770.1 | - | 6 | 1510 | |||
| BC098494.1 | - | 7 | 1518 | 6431607 | ||

Tfec
Genomic Map