siRNA Details
| Gene Symbol: Zfp275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160229.1 | NP_001153701.1 | 5 | 571 | ||
| NM_031494.2 | NP_113682.2 | 6 | 664 | |||
| GenBank mRNA | AB221659.1 | - | 3 | 379 | ||
| AK142957.1 | - | 5 | 571 | |||
| BC138002.1 | - | 6 | 569 | 8861021 | ||
| BC138003.1 | - | 6 | 569 | 8861022 | ||
| BY010614.1 | - | 2 | 215 | |||

Zfp275
Genomic Map