siRNA Details
| Gene Symbol: Zfp275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160229.1 | NP_001153701.1 | 5 | 3029 | ||
| NM_031494.2 | NP_113682.2 | 6 | 3122 | |||
| GenBank mRNA | AK090015.1 | - | 1 | 2291 | ||
| BC138002.1 | - | 6 | 3027 | 8861021 | ||
| BC138003.1 | - | 6 | 3027 | 8861022 | ||

Zfp275
Genomic Map