siRNA Details
| Gene Symbol: Abcg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031884.1 | NP_114090.1 | 4, 5 | 562 | ||
| GenBank mRNA | AF312713.2 | - | 4, 5 | 650 | ||
| AK149569.1 | - | 4, 5 | 533 | |||
| AY195872.1 | - | 4, 5 | 650 | |||
| AY195873.1 | - | 4, 5 | 650 | |||
| BC106766.2 | - | 4, 5 | 539 | 40039484 | ||

Abcg5
Genomic Map