siRNA Details
| Gene Symbol: Def8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253783.1 | NP_001240712.1 | 14 | 3240 | ||
| NM_001253784.1 | NP_001240713.1 | 13 | 3218 | |||
| NM_001281803.1 | NP_001268732.1 | 12 | 3222 | |||
| NM_054046.5 | NP_473387.1 | 12 | 3131 | |||
| GenBank mRNA | AK046271.1 | - | 12 | 3092 | ||
| AK050405.1 | - | 13 | 3189 | |||
| AK081295.1 | - | 12 | 3125 | |||
| AK087778.1 | - | 12 | 3103 | |||
| AK129019.1 | - | 12 | 3173 | |||
| AK133979.1 | - | 12 | 3109 | |||
| AK150334.1 | - | 12 | 3067 | |||
| AK150994.1 | - | 12 | 3067 | |||
| BC003306.1 | - | 14 | 3191 | 3154712 | ||

Def8
Genomic Map