siRNA Details
| Gene Symbol: Zfand3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148926.2 | NP_683728.1 | 5 | 2490 | ||
| GenBank mRNA | AK082915.1 | - | 4 | 2486 | ||
| AK148316.1 | - | 2520 | ||||
| BC024483.1 | - | 4 | 2152 | |||
| BC027161.1 | - | 4 | 2289 | |||
| BC034396.1 | - | 5 | 2411 | 3707476 | ||
| BC083124.1 | - | 5 | 2490 | 6334368 | ||

Zfand3
Genomic Map