siRNA Details
| Gene Symbol: Zfp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172462.4 | NP_766050.1 | 3 | 2053 | ||
| GenBank mRNA | AK032872.1 | - | 3 | 2068 | ||
| AK085077.1 | - | 2 | 2039 | |||
| BC058553.1 | - | 3 | 2051 | |||
| BC094590.1 | - | 2 | 2038 | 30616053 | ||
| BC139312.1 | - | 2 | 2033 | 8862334 | ||
| BC139315.1 | - | 2 | 2033 | 8862337 | ||

Zfp11
Genomic Map