siRNA Details
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009316.1 | NP_033342.1 | 3, 4 | 447 | ||
| NM_172688.3 | NP_766276.1 | 3, 4 | 447 | |||
| GenBank mRNA | AK046565.1 | - | 3, 4 | 413 | ||
| AK053911.1 | - | 3, 4 | 433 | |||
| AK137046.1 | - | 3, 4 | 448 | |||
| AK145302.1 | - | 3, 4 | 458 | |||
| AK159277.1 | - | 3, 4 | 411 | |||
| AK211827.1 | - | 1, 2 | 45 | |||
| BC006665.1 | - | 3, 4 | 438 | 3499247 | ||
| D76446.1 | - | 3, 4 | 447 | |||

Map3k7
Genomic Map