siRNA Details
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009316.1 | NP_033342.1 | 17 | 1903 | ||
| NM_172688.3 | NP_766276.1 | 16 | 1822 | |||
| GenBank mRNA | AK046565.1 | - | 16 | 1788 | ||
| AK053911.1 | - | 16 | 1808 | |||
| AK133642.1 | - | 10 | 949 | |||
| AK137046.1 | - | 16 | 1823 | |||
| AK159277.1 | - | 16 | 1786 | |||
| BC006665.1 | - | 17 | 1894 | 3499247 | ||
| D76446.1 | - | 16 | 1822 | |||

Map3k7
Genomic Map