siRNA Details
| Gene Symbol: Slc23a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194333.3 | NP_919314.2 | 3 | 439 | ||
| GenBank mRNA | BC094893.1 | - | 3 | 443 | 30017276 | |
| BC117790.1 | - | 3 | 426 | 40092152 | ||
| BC117791.1 | - | 3 | 426 | 40092153 | ||
| U25739.1 | - | 3 | 451 | |||

Slc23a3
Genomic Map