siRNA Details
| Gene Symbol: Esr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012754.1 | NP_036886.1 | 9 | 2217 | ||
| XM_006240221.2 | XP_006240283.1 | 10 | 2512 | |||
| GenBank mRNA | AB190769.1 | - | 8 | 2040 | ||
| AB190770.1 | - | 9 | 2094 | |||
| U57439.1 | - | 9 | 2217 | |||

Esr2
Genomic Map