siRNA Details
| Gene Symbol: Smad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013130.2 | NP_037262.2 | 7 | 2046 | ||
| XM_006255394.1 | XP_006255456.1 | 7 | 2192 | |||
| XM_006255395.2 | XP_006255457.1 | 7 | 2851 | |||
| XM_006255396.2 | XP_006255458.1 | 7 | 2082 | |||
| XM_006255397.1 | XP_006255459.1 | 7 | 2171 | |||
| GenBank mRNA | BC061757.1 | - | 7 | 2046 | 5599977 | |

Smad1
Genomic Map