siRNA Details
| Gene Symbol: Vdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017058.1 | NP_058754.1 | 9 | 1056 | ||
| GenBank mRNA | J04147.1 | - | 9 | 1056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vdr| Gene Symbol: Vdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017058.1 | NP_058754.1 | 9 | 1056 | ||
| GenBank mRNA | J04147.1 | - | 9 | 1056 | ||