siRNA Details
| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017327.1 | NP_059023.1 | 8, 9 | 1800 | ||
| GenBank mRNA | M12671.1 | - | 7, 8 | 964 | ||
| M17526.1 | - | 8, 9 | 1800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnao1| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017327.1 | NP_059023.1 | 8, 9 | 1800 | ||
| GenBank mRNA | M12671.1 | - | 7, 8 | 964 | ||
| M17526.1 | - | 8, 9 | 1800 | |||