siRNA Details
| Gene Symbol: Fasn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017332.1 | NP_059028.1 | 41 | 7170 | ||
| GenBank mRNA | J03514.1 | - | 5 | 839 | ||
| M76767.1 | - | 41 | 7170 | |||
| X13415.1 | - | 39 | 6968 | |||
| X62888.1 | - | 41 | 7175 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fasn| Gene Symbol: Fasn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017332.1 | NP_059028.1 | 41 | 7170 | ||
| GenBank mRNA | J03514.1 | - | 5 | 839 | ||
| M76767.1 | - | 41 | 7170 | |||
| X13415.1 | - | 39 | 6968 | |||
| X62888.1 | - | 41 | 7175 | |||