siRNA Details
| Gene Symbol: Pla2g2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019202.2 | NP_062075.1 | 6 | 1499 | ||
| XM_006239142.2 | XP_006239204.1 | 6 | 1517 | |||
| XM_006239143.2 | XP_006239205.1 | 5 | 1278 | |||
| XM_008764212.1 | XP_008762434.1 | 6 | 1592 | |||
| XM_008764213.1 | XP_008762435.1 | 6 | 1370 | |||
| XM_008764214.1 | XP_008762436.1 | 6 | 3041 | |||
| GenBank mRNA | BC097325.1 | - | 6 | 1499 | 7456563 | |
| U07798.1 | - | 6 | 1827 | |||

Pla2g2c
Genomic Map