siRNA Details
| Gene Symbol: Pla2g2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239142.2 | XP_006239204.1 | 6 | 3656 | ||
| XM_006239143.2 | XP_006239205.1 | 5 | 3417 | |||
| XM_008764212.1 | XP_008762434.1 | 6 | 3731 | |||
| XM_008764213.1 | XP_008762435.1 | 6 | 3509 | |||
| XM_008764214.1 | XP_008762436.1 | 6 | 5180 | |||
| GenBank mRNA | U07798.1 | - | 6 | 3966 | ||

Pla2g2c
Genomic Map