siRNA Details
| Gene Symbol: Stk39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019362.1 | NP_062235.1 | 18 | 2352 | ||
| XM_008761922.1 | XP_008760144.1 | 18 | 2096 | |||
| GenBank mRNA | AF099990.1 | - | 19 | 2057 | ||
| D88190.1 | - | 18 | 2352 | |||

Stk39
Genomic Map