siRNA Details
| Gene Symbol: Adam15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020308.1 | NP_064704.1 | 21 | 2628 | ||
| XM_006232742.2 | XP_006232804.1 | 23 | 2794 | |||
| XM_006232744.2 | XP_006232806.1 | 22 | 2722 | |||
| XM_008761203.1 | XP_008759425.1 | 23 | 2827 | |||
| XM_008761204.1 | XP_008759426.1 | 22 | 2755 | |||
| XM_008761205.1 | XP_008759427.1 | 21 | 2683 | |||
| GenBank mRNA | AJ251198.1 | - | 21 | 2628 | ||
| BC061796.1 | - | 23 | 2722 | 5597908 | ||

Adam15
Genomic Map