siRNA Details
| Gene Symbol: Isl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020471.1 | NP_065204.1 | 6 | 1245 | ||
| XM_006243089.1 | XP_006243151.1 | 6 | 1331 | |||
| GenBank mRNA | L35571.1 | - | 6 | 1245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isl2| Gene Symbol: Isl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020471.1 | NP_065204.1 | 6 | 1245 | ||
| XM_006243089.1 | XP_006243151.1 | 6 | 1331 | |||
| GenBank mRNA | L35571.1 | - | 6 | 1245 | ||