siRNA Details
| Gene Symbol: Mpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021265.3 | NP_067088.2 | 7 | 647 | ||
| XM_006245011.2 | XP_006245073.1 | 8 | 1934 | |||
| XM_008767130.1 | XP_008765352.1 | 8 | 1934 | |||
| XM_008767131.1 | XP_008765353.1 | 9 | 1074 | |||
| XM_008767132.1 | XP_008765354.1 | 10 | 960 | |||
| XM_008767133.1 | XP_008765355.1 | 6 | 529 | |||
| GenBank mRNA | AB030499.1 | - | 5 | 596 | ||
| AB030500.1 | - | 5 | 596 | |||
| AB030501.1 | - | 5 | 596 | |||

Mpp4
Genomic Map