siRNA Details
| Gene Symbol: Nucb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021663.2 | NP_067695.1 | 14 | 1588 | ||
| XM_006230095.2 | XP_006230157.1 | 13 | 1496 | |||
| GenBank mRNA | BC061778.1 | - | 14 | 1588 | 5621784 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nucb2| Gene Symbol: Nucb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021663.2 | NP_067695.1 | 14 | 1588 | ||
| XM_006230095.2 | XP_006230157.1 | 13 | 1496 | |||
| GenBank mRNA | BC061778.1 | - | 14 | 1588 | 5621784 | |