siRNA Details
| Gene Symbol: Trdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021666.2 | NP_067698.1 | 244 | |||
| GenBank mRNA | AF220558.1 | - | 426 | |||
| AF220559.1 | - | 426 | ||||
| AJ243303.1 | - | 329 | ||||
| AJ243304.2 | - | 244 | ||||
| AJ812275.1 | - | 329 | ||||
| AJ812276.1 | - | 329 | ||||
| FQ216749.1 | - | 335 | ||||
| FQ217225.1 | - | 2 | 179 | |||

Trdn
Genomic Map