siRNA Details
| Gene Symbol: Csad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134454.1 | NP_001127926.1 | 3, 4 | 354 | ||
| NM_021750.2 | NP_068518.1 | 4, 5 | 365 | |||
| XM_006242438.2 | XP_006242500.1 | 4, 5 | 774 | |||
| XM_006242439.1 | XP_006242501.1 | 4, 5 | 371 | |||
| XM_006242440.2 | XP_006242502.1 | 5, 6 | 850 | |||
| XM_006242441.2 | XP_006242503.1 | 4, 5 | 432 | |||
| XM_006242442.2 | XP_006242504.1 | 5, 6 | 465 | |||
| XM_008765782.1 | XP_008764004.1 | 4, 5 | 552 | |||
| GenBank mRNA | AJ132661.1 | - | 5, 6 | 470 | ||
| BC081804.1 | - | 4, 5 | 365 | 7106840 | ||
| EU786150.1 | - | 3, 4 | 337 | |||
| EU906909.1 | - | 3, 4 | 354 | |||
| M64755.1 | - | 4, 5 | 404 | |||
| U74492.1 | - | 2 | 215 | |||
| X94152.1 | - | 4, 5 | 523 | |||

Csad
Genomic Map