siRNA Details
| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021757.2 | NP_068525.1 | 3 | 242 | ||
| GenBank mRNA | AF128241.1 | - | 2 | 149 | ||
| BC087742.1 | - | 3 | 242 | 7314208 | ||
| FQ215141.1 | - | 3 | 262 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plrg1| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021757.2 | NP_068525.1 | 3 | 242 | ||
| GenBank mRNA | AF128241.1 | - | 2 | 149 | ||
| BC087742.1 | - | 3 | 242 | 7314208 | ||
| FQ215141.1 | - | 3 | 262 | |||