siRNA Details
Gene Symbol: Glp2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021848.1 | NP_068620.1 | 9 | 1016 | ||
GenBank mRNA | AF105368.1 | - | 9 | 1016 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
