siRNA Details
| Gene Symbol: Bcl2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021850.2 | NP_068622.1 | 3 | 575 | ||
| XM_006252016.2 | XP_006252078.1 | 3 | 765 | |||
| XM_008770692.1 | XP_008768914.1 | 2 | 1046 | |||
| XM_008770693.1 | XP_008768915.1 | 2 | 1122 | |||
| XM_008770694.1 | XP_008768916.1 | 3 | 1185 | |||
| GenBank mRNA | AF096291.1 | - | 1 | 377 | ||
| AY185098.1 | - | 3 | 546 | |||
| AY185099.1 | - | 3 | 280 | |||
| AY185100.1 | - | 1 | 885 | |||
| BC074021.1 | - | 3 | 575 | 7105621 | ||
| FQ212018.1 | - | 3 | 732 | |||

Bcl2l2
Genomic Map