siRNA Details
| Gene Symbol: Grm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022202.1 | NP_071538.1 | 1 | 326 | ||
| GenBank mRNA | U63288.1 | - | 1 | 326 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grm8| Gene Symbol: Grm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022202.1 | NP_071538.1 | 1 | 326 | ||
| GenBank mRNA | U63288.1 | - | 1 | 326 | ||
Set Membership: |
|