siRNA Details
| Gene Symbol: Unc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022206.1 | NP_071542.1 | 3 | 317 | ||
| XM_008771505.1 | XP_008769727.1 | 3 | 556 | |||
| GenBank mRNA | U87305.1 | - | 3 | 317 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Unc5a
Unc5a| Gene Symbol: Unc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022206.1 | NP_071542.1 | 3 | 317 | ||
| XM_008771505.1 | XP_008769727.1 | 3 | 556 | |||
| GenBank mRNA | U87305.1 | - | 3 | 317 | ||
 Back To Top
 Back To Top