siRNA Details
| Gene Symbol: Fut4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022219.3 | NP_071555.3 | 1 | 1037 | ||
| GenBank mRNA | AB049938.1 | - | 1 | 682 | ||
| BC061776.1 | - | 1 | 1037 | 5621698 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fut4| Gene Symbol: Fut4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022219.3 | NP_071555.3 | 1 | 1037 | ||
| GenBank mRNA | AB049938.1 | - | 1 | 682 | ||
| BC061776.1 | - | 1 | 1037 | 5621698 | ||