siRNA Details
| Gene Symbol: Mmp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022221.1 | NP_071557.1 | 2 | 197 | ||
| XM_006242496.2 | XP_006242558.1 | 2 | 278 | |||
| GenBank mRNA | AJ007288.1 | - | 2 | 197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp8| Gene Symbol: Mmp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022221.1 | NP_071557.1 | 2 | 197 | ||
| XM_006242496.2 | XP_006242558.1 | 2 | 278 | |||
| GenBank mRNA | AJ007288.1 | - | 2 | 197 | ||